Thermo Scientific Chemicals
Hydrochloric acid, ACS, HCl 36.5-38.0%, Thermo Scientific Chemicals
Catalog number: 033257.P6
6 lb., Each
Thermo Scientific Chemicals
Hydrochloric acid, ACS, HCl 36.5-38.0%, Thermo Scientific Chemicals
Catalog number: 033257.P6
6 lb., Each
Quantity
Catalog number: 033257.P6
also known as 033257-P6
Price (USD)
Quantity
-
Have Questions?
Description
Acidizing petroleum wells, chemical intermediate, ore reduction, food processing, pickling and metal cleaning, general cleaning, and laboratory reagent.Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.