Hydrochloric acid, ACS, HCl 36.5-38.0%, Thermo Scientific Chemicals
Hydrochloric acid, ACS, HCl 36.5-38.0%, Thermo Scientific Chemicals
Thermo Scientific™

Hydrochloric acid, ACS, HCl 36.5-38.0%, Thermo Scientific Chemicals

 
Have Questions?
Change viewbuttonViewtableView
Quantity:
6 x 6 lb.
6 lb.
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 033257.Q9
also known as 033257-Q9
Price (USD)/ Each
373.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
439.00 
Save 65.35 (15%)
-
Add to cart
Quantity:
6 x 6 lb.
Request bulk or custom format
Price (USD)/ Each
373.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
439.00 
Save 65.35 (15%)
Add to cart
Hydrochloric acid, ACS, HCl 36.5-38.0%, Thermo Scientific Chemicals
Catalog number033257.Q9
Price (USD)/ Each
373.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
439.00 
Save 65.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)10
Assay (unspecified)36.5-38.0% HCl
Heavy Metals(ICP-OES) 1ppm
Impurity contentFe 0.2ppm
Impurity contentNH₄ 3ppm
View more
Acidizing petroleum wells, chemical intermediate, ore reduction, food processing, pickling and metal cleaning, general cleaning, and laboratory reagent.Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
RUO – Research Use Only