Thermo Scientific™

Hydrofluoric acid, ACS, 48-51%, Thermo Scientific™

Catalog number: 033258.LB2
470 mL, Each
Thermo Scientific™

Hydrofluoric acid, ACS, 48-51%, Thermo Scientific™

Catalog number: 033258.LB2
470 mL, Each
Quantity
Catalog number: 033258.LB2
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7664-39-3
IUPAC Name
hydrogen fluoride
Molecular Formula
FH
InChI Key
KRHYYFGTRYWZRS-UHFFFAOYSA-N
SMILES
F
Trace Metals
(as Pb) 0.5ppm
Residue on ignition
5ppm
Impurity content
Fe 1ppm
Impurity content
PO₄ 1ppm
Impurity content
Sulfate and sulfite (as SO₄) 5ppm

Description

Used in aluminum production, fluorocarbons, pickling stainless steel, etching glass, acidizing oil wells, gasoline production and processing uraniumHydrofluoricacid is used in the glass etching, electronic and chemical industries. Hydrogen fluoride is mainly used in the production of organofluorine compounds, aluminum and chlorofluorocarbons. Hydrogen fluoride is also used for separating uranium isotopes, as a catalyst in the petroleum industry, and in stainless steel pickling. Hydrofluoric acid is used as a pickling agent and used in the semiconductor materials.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text