Thermo Scientific Chemicals

Nitric acid, ACS, fuming, 90%, Thermo Scientific Chemicals

Catalog number: 033261.K4
2.5 L, Each
Thermo Scientific Chemicals

Nitric acid, ACS, fuming, 90%, Thermo Scientific Chemicals

Catalog number: 033261.K4
2.5 L, Each
Quantity
Catalog number: 033261.K4
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7697-37-2
IUPAC Name
nitric acid
Molecular Formula
HNO3
InChI Key
GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES
O[N+]([O-])=O
Residue on ignition
0.002%
Impurity content
Cl 0.7ppm
Trace Metals
(as Pb) 5ppm
Assay (unspecified)
90% HNO₃ min
Comment
Dilution test P.T.

Description

Main commercial usage of Nitric acid, ACS, fuming, 90% is in the production of fertilizers and reagents for nitration. It is used in the applications for the production of explosives, nylon precursors and specialty organic compounds. Nitric acid is indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions. It is used as a chemical doping agent for organic semiconductors and in purification processes for raw carbon nanotubes, artificially age pine and maple.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text