Thermo Scientific Chemicals

Silicic acid hydrate, Thermo Scientific Chemicals

Catalog number: 033270.22
100 g, Each
Thermo Scientific Chemicals

Silicic acid hydrate, Thermo Scientific Chemicals

Catalog number: 033270.22
100 g, Each
Quantity
Catalog number: 033270.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1343-98-2
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Appearance (Color)
White
Form
Powder, -100 mesh
Loss on ignition
40-70%

Description

Silicic acid hydrate is used for studying neurodegenerative diseases due to its ability to inhibit aluminum uptake. It is utilized for the manufacture catalyst and catalyst carrier. It is involved in tungsten filament production as chemical reagent and flux. Further, it is used for oil and wax decoloring.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text