Thermo Scientific Chemicals

Succinic acid, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 033272.A3
2 kg, Each
Thermo Scientific Chemicals

Succinic acid, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 033272.A3
2 kg, Each
Quantity
Catalog number: 033272.A3
also known as 033272-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-15-6
IUPAC Name
butanedioic acid
Molecular Formula
C4H6O4
InChI Key
KDYFGRWQOYBRFD-UHFFFAOYSA-N
SMILES
OC(=O)CCC(O)=O
Chloride content
0.001%, max
Heavy Metals (as Pb)
5ppm, max
Appearance (Color)
White to pale cream
Form
crystalline powder
Residue on ignition
0.02%, max

Description

Succinic acid acts as a leaching agent in extracting lithium, cobalt from used Li-ion batteries and magnesium from magnesite ore. It is used as a starting material in the synthesis of new elastic polyesters, fumaric acid, succinic anhydride and alkyd resins. It plays an important role as an acidity regulator in the food and beverage industry; as an excipient in pharmaceutical products and acts as a cocrystallising agent in organic synthesis. Its derivative viz diethyl ester is used as a substrate in the Stobbe condensation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text