Thermo Scientific Chemicals

Sulfurous acid, ACS, 6.0% SO{2} min, Thermo Scientific Chemicals

Catalog number: 033275.AY
4 x 500 mL, Each
Thermo Scientific Chemicals

Sulfurous acid, ACS, 6.0% SO{2} min, Thermo Scientific Chemicals

Catalog number: 033275.AY
4 x 500 mL, Each
Quantity
Catalog number: 033275.AY
also known as 033275-AY
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7782-99-2
IUPAC Name
sulfurous acid
Molecular Formula
H2O3S
InChI Key
LSNNMFCWUKXFEE-UHFFFAOYSA-N
SMILES
OS(O)=O
Impurity content
Cl 5ppm
Trace Metals
(as Pb) 2ppm
Residue on ignition
0.005%
Assay (unspecified)
6.0% SO₂ min
Impurity content
Fe 5ppm

Description

Sulfurous acid is used as a reducing agent, bleaching agent and disinfectant. It is utilized for bleaching of moistened wool, straw for hats, and other materials injured by chlorine. It finds application as a disinfectant as well as used externally for its parasiticidal effect in various skin diseases. Its derivative viz. sulfites are included in antiseptics, antifermentatives, and antizymotics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text