Thermo Scientific Chemicals

Dithizone, ACS, 85+%, Thermo Scientific Chemicals

Catalog number: 033283.18
50 g, Each
Thermo Scientific Chemicals

Dithizone, ACS, 85+%, Thermo Scientific Chemicals

Catalog number: 033283.18
50 g, Each
Quantity
Catalog number: 033283.18
also known as 033283-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60-10-6
IUPAC Name
3-(phenylamino)-1-(phenylimino)thiourea
Molecular Formula
C13H12N4S
InChI Key
UOFGSWVZMUXXIY-UHFFFAOYSA-N
SMILES
S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
Comment
Ratio of absorbance 1.55
Trace Metals
(as Pb) 0.002%
Residue on ignition
0.3%

Description

Dithizone is used as an indicator for minute amounts of lead. It acts as a chelating agent for heavy metal poisoning. As a ligand, it forms complexes with metals like zinc, lead and mercury. Further, it is used to assess the purity of human pancreatic islet preparations, which is used for transplantation in patients with type 1 diabetes. In addition to this, it is used as a titrant in the quantitative determination of heavy metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text