Thermo Scientific Chemicals

Calcium carbonate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 033295.22
100 g, Each
Thermo Scientific Chemicals

Calcium carbonate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 033295.22
100 g, Each
Quantity
Catalog number: 033295.22
also known as 033295-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
471-34-1
IUPAC Name
calcium carbonate
Molecular Formula
CCaO3
InChI Key
VTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES
[Ca++].[O-]C([O-])=O
Impurity content
Na 0.1%
Impurity content
Cl 0.001%
Impurity content
F 0.0015%
Impurity content
Insoluble in dilute hydrochloric acid 0.01%
Impurity content
Mg 0.02%

Description

Calcium carbonate is used medicinally as a calcium supplement or as an antacid. It is also used for industrial purposes. Calcium carbonate is the most widely used mineral in the paper, plastics, paints and coatings industries. Hierarchical hollow calcium carbonate particles show a useful application as an anticancer drug carrier. It plays an essential role to catalyze alcoholysis of fats and oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text