Thermo Scientific Chemicals

Calcium oxide, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 033299.36
500 g, Each
Thermo Scientific Chemicals

Calcium oxide, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 033299.36
500 g, Each
Quantity
Catalog number: 033299.36
also known as 033299-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1305-78-8
IUPAC Name
calcium oxidandiide
Molecular Formula
CaO
InChI Key
BRPQOXSCLDDYGP-UHFFFAOYSA-N
SMILES
[O--].[Ca++]
Loss on ignition
5% max
Insoluble material
1.5% max in CH₃COOH and NH₄OH
Trace Metals
(as Pb) 0.01% max

Description

Polyethylene oxide is used as lubricants for rubber molds, textiles, metalworking and ceramics production. Further, it is used in food packaging, water-based paints, polishes, paper coatings, dispersing agents and chemical intermediates. In addition, it is used to impart viscosity to and modify the flow of aqueous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text