Calcium sulfate dihydrate, 99%, Thermo Scientific Chemicals
Calcium sulfate dihydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium sulfate dihydrate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 033301.A1
also known as 033301-A1
Price (USD)/ Each
74.65
Online exclusive
82.90 
Save 8.25 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
74.65
Online exclusive
82.90 
Save 8.25 (10%)
Add to cart
Calcium sulfate dihydrate, 99%, Thermo Scientific Chemicals
Catalog number033301.A1
Price (USD)/ Each
74.65
Online exclusive
82.90 
Save 8.25 (10%)
-
Add to cart
Chemical Identifiers
CAS10101-41-4
IUPAC Namecalcium dihydrate sulfate
Molecular FormulaCaH4O6S
InChI KeyPASHVRUKOFIRIK-UHFFFAOYSA-L
SMILESO.O.[Ca++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
Used as portland-cement retarder, tile and plaster. It is a source of sulfur and sulfuric acid. Also used in polishing powder and paints, dyeing and calico printing. It is likewise employed in zinc metallurgy. It is employed in drying industrial gases, solids and many organic liquids. In granulated form it is used as soil conditioner. Used for food additives in the soy manufacturing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only