Thermo Scientific Chemicals

Chromium(III) sulfate hydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 033303.36
500 g, Each
Thermo Scientific Chemicals

Chromium(III) sulfate hydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 033303.36
500 g, Each
Quantity
Catalog number: 033303.36
also known as 033303-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15244-38-9
IUPAC Name
dichromium(3+) trisulfate
Molecular Formula
Cr2O12S3
InChI Key
GRWVQDDAKZFPFI-UHFFFAOYSA-H
SMILES
[Cr+3].[Cr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Form
Crystals or crystalline powder
Assay (special)
Chromium (Cr): 18.0-26.0%
Appearance (Color)
Green to green-black

Description

Chromium(III) sulfate hydrate is widely used in making chrome tanned leather, paints, inks and ceramics. It is also used in preparation of chromium metal, dye, in printing and dyeing industry. It is involved in the preparation of chrome catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text