Thermo Scientific Chemicals

Copper(II) carbonate dihydroxide, Cu 55% min, Thermo Scientific Chemicals

Catalog number: 033305.A3
2 kg, Each
Thermo Scientific Chemicals

Copper(II) carbonate dihydroxide, Cu 55% min, Thermo Scientific Chemicals

Catalog number: 033305.A3
2 kg, Each
Quantity
Catalog number: 033305.A3
also known as 033305-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12069-69-1
IUPAC Name
dicopper(2+) dihydroxide carbonate
Molecular Formula
CH2Cu2O5
InChI Key
ZMMDPCMYTCRWFF-UHFFFAOYSA-J
SMILES
[OH-].[OH-].[Cu++].[Cu++].[O-]C([O-])=O
Assay (unspecified)
95% min

Description

Copper(II) carbonate dihydroxide is used as pigments and in treatment for copper deficiency in ruminants. It is also used in pyrotechnics, in sweetening of petrol sour crude stock and animal and poultry feeds. It finds application in some types of make-up such as lipstick. In analytical chemistry, it is used as a reagent for analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text