Thermo Scientific Chemicals

Cyclohexanone, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 033309.K2
1 L, Each
Thermo Scientific Chemicals

Cyclohexanone, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 033309.K2
1 L, Each
Quantity
Catalog number: 033309.K2
also known as 033309-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-94-1
IUPAC Name
cyclohexanone
Molecular Formula
C6H10O
InChI Key
JHIVVAPYMSGYDF-UHFFFAOYSA-N
SMILES
O=C1CCCCC1
Color (APHA)
≤10
Assay (GC)
≥99.0%
Residual water
≤0.05%
Residue on Evaporation
≤0.05%

Description

Cyclohexanone is used as an industrial solvent and paint remover. It acts as a precursor to nylon 6,6 and nylon 6 and cyclohexanone oxime, which gives caprolactam on rearrangement. Further, it is used as a chemical reaction medium, adhesives, sealants and agricultural products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text