Thermo Scientific Chemicals

Dimethylglyoxime, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 033310.18
50 g, Each
Thermo Scientific Chemicals

Dimethylglyoxime, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 033310.18
50 g, Each
Quantity
Catalog number: 033310.18
also known as 033310-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-45-4
IUPAC Name
(E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine
Molecular Formula
C4H8N2O2
InChI Key
JGUQDUKBUKFFRO-GGWOSOGESA-N
SMILES
C\C(=N/O)\C(\C)=N\O
Assay (unspecified)
≥99.0%
Comment
Suitability for nickel determination: Passes Test
Residue on ignition
≤0.05%
Impurity content
Insoluble in ethanol: ≤0.05%
Melting Point
ca 240°C

Description

Dimethylglyoxime, is used as a chelating agent which forms complexes with metals such as copper, nickel, and palladium. Dimethylglyoxime reacts with a nickel salt in a green solution to generate a red colored insoluble tetradentate coordinate complex of nickel. It is used as a reagent for the colorimetric determination of urea and ureido compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text