Formaldehyde, 37% in aq. soln., ACS, 36.5-38.0%, stab. with 10-15% methanol
Formaldehyde, 37% in aq. soln., ACS, 36.5-38.0%, stab. with 10-15% methanol
Thermo Scientific Chemicals

Formaldehyde, 37% in aq. soln., ACS, 36.5-38.0%, stab. with 10-15% methanol

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
4 x 1 L
4 L
4 x 4 L
Catalog number 033314.K2
also known as 033314-K2
Price (USD)/ Each
55.65
Online exclusive
62.10 
Save 6.45 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
55.65
Online exclusive
62.10 
Save 6.45 (10%)
Add to cart
Formaldehyde, 37% in aq. soln., ACS, 36.5-38.0%, stab. with 10-15% methanol
Catalog number033314.K2
Price (USD)/ Each
55.65
Online exclusive
62.10 
Save 6.45 (10%)
-
Add to cart
Chemical Identifiers
CAS50-00-0
IUPAC Nameformaldehyde
Molecular FormulaCH2O
InChI KeyWSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILESC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)≤10
Free acid (titration)≤0.006meq/g
Impurity contentSO₄: ≤ 0.002%
Appearance (Color)Colorless
FormLiquid
View more
Formaldehyde is used as a denaturant in formaldehyde-agarose gel electrophoresis of RNA. It is also used in fiber board, plywood, cigarette smoke, fuel burning appliances, kerosene space heaters and in photography. It acts as a precursor in the manufacturing of automobiles and is used to make components for transmissions, electrical systems, engine blocks, door panels, axles and brake shoes. It serves as a disinfectant. It is also an ingredient in glues and is used as a preservative in medical laboratories, as an embalming fluid and as a sterilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only