Thermo Scientific Chemicals

Iron(III) sulfate hydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 033316.A1
1 kg, Each
Thermo Scientific Chemicals

Iron(III) sulfate hydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 033316.A1
1 kg, Each
Quantity
Catalog number: 033316.A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15244-10-7
IUPAC Name
diiron(3+) hydrate trisulfate
Molecular Formula
Fe2H2O13S3
InChI Key
VXWSFRMTBJZULV-UHFFFAOYSA-H
SMILES
O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Assay (unspecified)
73% Fe₂(SO₄)₃ min

Description

Iron(III) sulfate is also used in pigments, pickling baths for aluminum and steel. It acts as a mordant in dyeing, as a coagulant for industrial wastes. In the medical field, it is used as an astringent and styptic. It is involved in the preparation of aromatic esters from the corresponding acids and alcohols. Further, it is employed in the oxidation of pyrite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text