Thermo Scientific™

Hexanes, mixed isomers, ACS, 98.5+%, Thermo Scientific Chemicals

Catalog number: 033321.M1
4 x 1 L, Each
Thermo Scientific™

Hexanes, mixed isomers, ACS, 98.5+%, Thermo Scientific Chemicals

Catalog number: 033321.M1
4 x 1 L, Each
Quantity
Catalog number: 033321.M1
also known as 033321-M1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
92112-69-1
IUPAC Name
hexane
Molecular Formula
C6H14
InChI Key
VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES
CCCCCC
Free acid (titration)
(Water soluble) ≤0.0003 meq/g
Form
Liquid
Color (APHA)
≤10
Assay (GC)
≥98.5% hexanes ( sum of 5 isomers, total hexanes, plus methylcyclopentane
Impurity content
Sulfur compounds (as S) ≤0.005%

Description

Hexane mixed isomers is used in the laboratory to extract oil and grease contaminants from water and soil for analysis. As a non-polar solvent, it is used in chromatography. It plays important role in the preparation of glues for shoes, leather products, and roofing. It is also used for reactions that involve very strong bases such as the preparation of Grignard reagents. Also involved in the preparation of organolithiums compounds namely butyl lithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text