Thermo Scientific Chemicals

Lead(II) carbonate, basic, ACS, Thermo Scientific Chemicals

Catalog number: 033328.30
250 g, Each
Thermo Scientific Chemicals

Lead(II) carbonate, basic, ACS, Thermo Scientific Chemicals

Catalog number: 033328.30
250 g, Each
Quantity
Catalog number: 033328.30
also known as 033328-30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1319-46-6
IUPAC Name
tris(λ²-lead(2+)) dihydroxide dicarbonate
Molecular Formula
C2H2O8Pb3
InChI Key
RYZCLUQMCYZBJQ-UHFFFAOYSA-H
SMILES
[OH-].[OH-].[Pb++].[Pb++].[Pb++].[O-]C([O-])=O.[O-]C([O-])=O
Impurity content
Cl 0.002%
Impurity content
Fe 0.005%
Impurity content
Insoluble in dilute acetic acid 0.02%
Impurity content
Nitrate and nitrite (as NO₃) P.T. (limit about 0.005%)
Impurity content
Cd 0.002%

Description

Lead(II) carbonate is used in ceramic glaze and exterior paint.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text