Thermo Scientific Chemicals

Lead(II) oxide, low silver, Thermo Scientific Chemicals

Catalog number: 033330.36
500 g, Each
Thermo Scientific Chemicals

Lead(II) oxide, low silver, Thermo Scientific Chemicals

Catalog number: 033330.36
500 g, Each
Quantity
Catalog number: 033330.36
also known as 033330-36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1317-36-8
IUPAC Name
plumbanone
Molecular Formula
OPb
InChI Key
YEXPOXQUZXUXJW-UHFFFAOYSA-N
SMILES
O=[Pb]
Form
Powder
Appearance (Color)
Yellow to yellow-orange to red
Assay from Supplier's CofA
≥99.9% (PbO)
Impurity content
Silver (Ag): ≤0.0015%

Description

Lead(II) oxide is used in storage batteries, ceramic cements and fluxes, pottery and glazes, in chromium pigments, in oil refining, in paints, and in the assay of precious metal ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • As storage batteries, ceramic cements and fluxes, pottery and glazes, in chromium pigments, in oil refining, in paints, and in the assay of precious metal ores
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text