Thermo Scientific Chemicals

Magnesium carbonate hydroxide tetrahydrate, White powder, MgO approx.40-43.5%, Thermo Scientific Chemicals

Catalog number: 033333.22
100 g, Each
Thermo Scientific Chemicals

Magnesium carbonate hydroxide tetrahydrate, White powder, MgO approx.40-43.5%, Thermo Scientific Chemicals

Catalog number: 033333.22
100 g, Each
Quantity
Catalog number: 033333.22
also known as 033333-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39409-82-0
IUPAC Name
pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
Molecular Formula
C4H10Mg5O18
InChI Key
YMKSTRLFWJQKFP-UHFFFAOYSA-D
SMILES
O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Assay (unspecified)
40-43.5% MgO

Description

Magnesium carbonate hydroxide tetrahydrate is used as a mixture with huntite and as filler in polymers. It acts as a flame retardant and fire retardant additive for polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text