Manganese(II) nitrate, 50% w/w aq. soln., Thermo Scientific Chemicals
Manganese(II) nitrate, 50% w/w aq. soln., Thermo Scientific Chemicals
Thermo Scientific™

Manganese(II) nitrate, 50% w/w aq. soln., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 033340.36
also known as 033340-36
Price (USD)/ Each
50.65
Online exclusive
56.10 
Save 5.45 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
50.65
Online exclusive
56.10 
Save 5.45 (10%)
Add to cart
Manganese(II) nitrate, 50% w/w aq. soln., Thermo Scientific Chemicals
Catalog number033340.36
Price (USD)/ Each
50.65
Online exclusive
56.10 
Save 5.45 (10%)
-
Add to cart
Chemical Identifiers
CAS10377-66-9
IUPAC Namemanganese(2+) dinitrate
Molecular FormulaMnN2O6
InChI KeyMIVBAHRSNUNMPP-UHFFFAOYSA-N
SMILES[Mn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)15.35-16.00% Mn
Impurity content0.001% Cl max
Impurity content0.001% Cu max
Impurity content0.002% Ni max
Impurity content0.01% Zn max
View more
Manganese(II) nitrate is used as a precursor to prepare the oxides of manganese. It is involved in the preparation of electronic components. Further, it is utilized for the phosphorous treatment of metal surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only