Thermo Scientific Chemicals

Manganese(II) nitrate, 50% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 033340.A3
2 kg, Each
Thermo Scientific Chemicals

Manganese(II) nitrate, 50% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 033340.A3
2 kg, Each
Quantity
Catalog number: 033340.A3
also known as 033340-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10377-66-9
IUPAC Name
manganese(2+) dinitrate
Molecular Formula
MnN2O6
InChI Key
MIVBAHRSNUNMPP-UHFFFAOYSA-N
SMILES
[Mn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Assay (unspecified)
15.35-16.00% Mn
Impurity content
0.001% Cl max
Impurity content
0.001% Cu max
Impurity content
0.002% Ni max
Impurity content
0.01% Zn max

Description

Manganese(II) nitrate is used as a precursor to prepare the oxides of manganese. It is involved in the preparation of electronic components. Further, it is utilized for the phosphorous treatment of metal surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text