Thermo Scientific Chemicals

D-Mannitol, ACS

Catalog number: 033342.A4
2.5 kg, Each
Thermo Scientific Chemicals

D-Mannitol, ACS

Catalog number: 033342.A4
2.5 kg, Each
Quantity
Catalog number: 033342.A4
also known as 033342-A4
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69-65-8
IUPAC Name
hexane-1,2,3,4,5,6-hexol
Molecular Formula
C6H14O6
InChI Key
FBPFZTCFMRRESA-UHFFFAOYNA-N
SMILES
OCC(O)C(O)C(O)C(O)CO
Impurity content
Reducing sugars P.T.
Free acid (titration)
0.0008meq/g
Optical Rotation
A25/D +23.3? to +24.3?
Insoluble material
0.01%
Trace Metals
(as Pb) 5ppm

Description

D-Mannitol is used with boric acid to make dry electrolytic condensers for radio applications, artificial resins and plasticizers. It is used as excipient for chewable tablets due to its pleasant taste. It is used in volumetric analysis of boric acid in analytical chemistry. It is also used in the food industry as anticaking, free-flow agent, flavoring agent, stabilizer, thickener and nutritive sweetener. Further, it is used in medicine as diuretic and in the determination of renal function. In addition, it is used as a formulation aid, lubricant, non-nutritive sweetener and solvent for foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text