Thermo Scientific Chemicals

Potassium hydrogen phthalate, primary standard, ACS, 99.95-100.05%, Thermo Scientific Chemicals

Catalog number: 033351.A1
1 kg, Each
Thermo Scientific Chemicals

Potassium hydrogen phthalate, primary standard, ACS, 99.95-100.05%, Thermo Scientific Chemicals

Catalog number: 033351.A1
1 kg, Each
Quantity
Catalog number: 033351.A1
also known as 033351-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
877-24-7
IUPAC Name
potassium 2-carboxybenzoate
Molecular Formula
C8H5KO4
InChI Key
IWZKICVEHNUQTL-UHFFFAOYSA-M
SMILES
[K+].OC(=O)C1=CC=CC=C1C([O-])=O
Form
Powder or crystals or crystalline powder
Heavy Metals
(as Pb): ≤5ppm
Impurity content
Fe: ≤5ppm
Impurity content
Sulfur Compounds (as S): ≤0.002%
pH
4.00-4.02 (0.05M solution, 25°C)

Description

As a primary standard for preparing volumetric alkali solutions and as a buffer in pH determinations. Potassium hydrogen phthalate is used as a primary standard for acid- base titrations as well as for calibrating pH meters. It acts as a buffer in pH determination. It finds application as a useful standard for total organic carbon testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text