Thermo Scientific Chemicals

Potassium hexacyanoferrate(II) trihydrate, ACS, 98.5-102.0%, Thermo Scientific Chemicals

Catalog number: 033358.22
100 g, Each
Thermo Scientific Chemicals

Potassium hexacyanoferrate(II) trihydrate, ACS, 98.5-102.0%, Thermo Scientific Chemicals

Catalog number: 033358.22
100 g, Each
Quantity
Catalog number: 033358.22
also known as 033358-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14459-95-1
IUPAC Name
tetrapotassium hexacyanoironbis(ylium) trihydrate
Molecular Formula
C6H6FeK4N6O3
InChI Key
NLYKFUFIUWPPIZ-UHFFFAOYSA-N
SMILES
O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
Impurity content
Cl 0.01%
Impurity content
SO₄ P.T. (limit about 0.01%)
Insoluble material
0.005%

Description

Potassium hexacyanoferrate(II) trihydrate is widely used in anticaking agent for both road and table salt. It is involved in the purification of tin and separation of copper from molybdenum ores. It plays a vital role in the in production of wine and citric acid. In analytical chemistry, it is used to determine the concentration of potassium permanganate. It is in combination with potassium ferricyanide and phosphate buffered solution acts as buffer for beta-galatosidase. Further, it is used as fertilizers for plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text