Thermo Scientific Chemicals

Ethanol, Alcohol Reagent, anhydrous, denatured, ACS, 94-96%, Thermo Scientific Chemicals

Catalog number: 033361.K2
1 L, Each
Thermo Scientific Chemicals

Ethanol, Alcohol Reagent, anhydrous, denatured, ACS, 94-96%, Thermo Scientific Chemicals

Catalog number: 033361.K2
1 L, Each
Quantity
Catalog number: 033361.K2
also known as 033361-K2
Price (USD)
Have Questions?

Chemical Identifiers

CAS
64-17-5
IUPAC Name
ethanol
Molecular Formula
C2H6O
InChI Key
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES
CCO
Appearance (Color)
Clear colorless
Assay (GC)
Within 94.0-96.0% (V/V) methanol and ethyl alcohol
Assay (GC)
Isopropyl alcohol (2-propanol)(v/v) 4.0-6.0%
Water Content (Karl Fischer Titration)
≤0.5%
Residue on Evaporation
≤0.001%

Description

Ethanol is used in antibacterial hand sanitizer gels, as an antidote to methanol poisoning and an antitussive agent. It is acting as a solvent to remove the paint and ink stain on curtains, dress and upholstery. It is utilized as a fuel in camping and primus stoves. It is likewise employed to clean the windows, manufacturing nail-polish and to test pectin levels in jam and jelly making. It finds application in personal care products such as mouthwashes, perfumes and deodorants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text