Thermo Scientific Chemicals

Sodium diethyldithiocarbamate trihydrate, ACS, Thermo Scientific Chemicals

Catalog number: 033379.36
500 g, Each
Thermo Scientific Chemicals

Sodium diethyldithiocarbamate trihydrate, ACS, Thermo Scientific Chemicals

Catalog number: 033379.36
500 g, Each
Quantity
Catalog number: 033379.36
also known as 033379-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20624-25-3
IUPAC Name
sodium (diethylcarbamothioyl)sulfanide trihydrate
Molecular Formula
C5H16NNaO3S2
InChI Key
WWGXHTXOZKVJDN-UHFFFAOYSA-M
SMILES
O.O.O.[Na+].CCN(CC)C([S-])=S
Comment
Sensitivity to copper P.T.
Impurity content
Sodium (as Na₂SO₄) 30.5-32.5%
Solution Test
in water P.T.

Description

Sodium diethyldithiocarbamate trihydrate is a spin trapping reagent used in conjunction with ferrous ion to detect nitric oxide in brain, kidney, liver and other tissues. It is used as sensitive reagent for the determination of copper, rubber vulcanization accelerator and finds application in the determination of other metals such as zinc, cobalt, platinum and palladium. Furthermore, it is an inhibitor of superoxide dismutase, ascorbate oxidase and other enzymes as well as inhibits the induction of macrophage nitric oxide synthase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text