Thermo Scientific Chemicals

Sodium hydrosulfite, tech., 85+%, Thermo Scientific Chemicals

Catalog number: 033381.22
100 g, Each
Thermo Scientific Chemicals

Sodium hydrosulfite, tech., 85+%, Thermo Scientific Chemicals

Catalog number: 033381.22
100 g, Each
Quantity
Catalog number: 033381.22
also known as 033381-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7775-14-6
IUPAC Name
disodium sulfinatosulfinate
Molecular Formula
Na2O4S2
InChI Key
JVBXVOWTABLYPX-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]S(=O)S([O-])=O
Assay (unspecified)
≥85.0%
Appearance (Color)
White
Form
Powder

Description

Sodium hydrosulfite is used as a reducing agent in aqueous solutions, sulfonating agent , chelating agent and decolorizing agent in organic reactions. It finds application in water treatment, gas purification, cleaning, leather, polymers, photography, and many others. It is involved in chemical enhanced oil recovery to stabilize polyacrylamide polymers against radical degradation in the presence of iron. It plays an important role to determine the iron content in soil chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text