Thermo Scientific Chemicals

Sodium perborate tetrahydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 033383.14
25 g, Each
Thermo Scientific Chemicals

Sodium perborate tetrahydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 033383.14
25 g, Each
Quantity
Catalog number: 033383.14
also known as 033383-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10486-00-7
IUPAC Name
disodium tetrahydroxy-1,2,4,5,3,6-tetraoxadiborinane-3,6-diuide hexahydrate
Molecular Formula
B2H16Na2O14
InChI Key
MCYPYXIRGFEFGL-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[Na+].[Na+].O[B-]1(O)OO[B-](O)(O)OO1
Assay (unspecified)
NaBO₃?4H₂O 96% min

Description

Sodium perborate tetrahydrate is mainly used in textile mills as a bleaching agent for a variety of fabrics, namely, wool, cotton, linen, rayon, etc, and as oxidizing and fixing agent for dyes. It is also used in dentifrices, denture cleaners and pharmaceuticals due to its cleansing, deodorizing and antiseptic properties and is incorporated in soaps and detergent powders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text