Thermo Scientific Chemicals

Sodium succinate hexahydrate, Thermo Scientific Chemicals

Catalog number: 033386.30
250 g, Each
Thermo Scientific Chemicals

Sodium succinate hexahydrate, Thermo Scientific Chemicals

Catalog number: 033386.30
250 g, Each
Quantity
Catalog number: 033386.30
also known as 033386-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6106-21-4
IUPAC Name
disodium butanedioate hexahydrate
Molecular Formula
C4H16Na2O10
InChI Key
ZBTUYCUNQBRXOR-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Na+].[Na+].[O-]C(=O)CCC([O-])=O
Appearance (Color)
White
pH
0.4M solution: 6.0-9.0
Assay from Supplier's CofA
≥97.0% to ≤102.0%
Heavy Metals (as Pb)
≤0.002% (20ppm)
Solution Test
Water, 10%: clear solution

Description

Sodium succinate hexahydrate is used as an ingredient of topical preparations for the treatment of cataract.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text