Thermo Scientific Chemicals

Strontium carbonate, 97.5%, Thermo Scientific Chemicals

Catalog number: 033392.30
250 g, Each
Thermo Scientific Chemicals

Strontium carbonate, 97.5%, Thermo Scientific Chemicals

Catalog number: 033392.30
250 g, Each
Quantity
Catalog number: 033392.30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1633-05-2
IUPAC Name
strontium(2+) carbonate
Molecular Formula
CO3Sr
InChI Key
LEDMRZGFZIAGGB-UHFFFAOYSA-L
SMILES
[Sr++].[O-]C([O-])=O
Assay (unspecified)
97.5% min

Description

As a catalyst, in radiation-resistant glass, in ceramic ferrites, and in pyrotechnicsUsed as a colorant in fireworks. It is also used in ceramics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text