Thermo Scientific Chemicals

Zinc sulfate heptahydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals

Catalog number: 033399.22
100 g, Each
Thermo Scientific Chemicals

Zinc sulfate heptahydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals

Catalog number: 033399.22
100 g, Each
Quantity
Catalog number: 033399.22
also known as 033399-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7446-20-0
IUPAC Name
zinc(2+) heptahydrate sulfate
Molecular Formula
H14O11SZn
InChI Key
RZLVQBNCHSJZPX-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
Assay (unspecified)
99.0-103.0% O₄SZn.7H₂O
Impurity content
Fe 0.001%
Impurity content
Mg 0.005%
Impurity content
Pb 0.003%
Impurity content
Ca 0.005%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text