Osmium(III) chloride trihydrate, Premion™, 99.99% (metals basis), Os 52-56%, Thermo Scientific Chemicals
Osmium(III) chloride trihydrate, Premion™, 99.99% (metals basis), Os 52-56%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Osmium(III) chloride trihydrate, Premion™, 99.99% (metals basis), Os 52-56%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
0.25 g
1 g
Catalog number 033480.06
also known as 033480-06
Price (USD)/ Each
767.65
Online exclusive
853.00 
Save 85.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
767.65
Online exclusive
853.00 
Save 85.35 (10%)
Add to cart
Osmium(III) chloride trihydrate, Premion™, 99.99% (metals basis), Os 52-56%, Thermo Scientific Chemicals
Catalog number033480.06
Price (USD)/ Each
767.65
Online exclusive
853.00 
Save 85.35 (10%)
-
Add to cart
Chemical Identifiers
CAS135296-80-9
IUPAC Nametrichloroosmium trihydrate
Molecular FormulaCl3H6O3Os
InChI KeyUACQLNPCDXDCID-UHFFFAOYSA-K
SMILESO.O.O.Cl[Os](Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Osmium(III) chloride trihydrate is used as a reagent for preparation of dichlorodihydridoosmium complexes. It is also useful as starting materials and an efficient alkane oxidation catalyst in the presence of pyridine and hydrogen oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only