Thermo Scientific Chemicals
Sodium bis(trimethylsilyl)amide, 98%, Thermo Scientific Chemicals
Catalog number: 033505.04
2 g, Each
Thermo Scientific Chemicals
Sodium bis(trimethylsilyl)amide, 98%, Thermo Scientific Chemicals
Catalog number: 033505.04
2 g, Each
Quantity
Catalog number: 033505.04
also known as 033505-04
Price (USD)
Quantity
-
Have Questions?
Description
Sodium bis(trimethylsilyl)amide is used as a base in organic synthesis as well as to deprotonate other compounds containing weakly acidic O-H, S-H, and N-H bonds. It is used to prepare amines by reacting with alkyl halides. It finds application in certain organic reactions viz. generation of carbenes by dehydrohalogenation of halocarbons and Wittig reagents by deprotonation of phosphonium salts. It accelerates the polymerization of pheneylacetylene in conjunction with rhodium(I) catalysis. Furthermore, it is used for the preparation of lanthanide complexes.