Aluminum, AAS standard solution, Specpure™ Al 1000μg/mL, Thermo Scientific Chemicals
Aluminum, AAS standard solution, Specpure™ Al 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum, AAS standard solution, Specpure™ Al 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 033557.AP
also known as 033557-AP
Price (USD)/ Each
68.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
80.50 
Save 11.85 (15%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
68.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
80.50 
Save 11.85 (15%)
Add to cart
Aluminum, AAS standard solution, Specpure™ Al 1000μg/mL, Thermo Scientific Chemicals
Catalog number033557.AP
Price (USD)/ Each
68.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
80.50 
Save 11.85 (15%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialAluminum
CAS11145-27-0
Health Hazard 1H290
Quantity500 mL
Health Hazard 3P234-P390
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only