This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 033559.AP
also known as 033559-AP
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
78.60
Save 11.95 (15%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
78.60
Save 11.95 (15%)
Add to cart
Arsenic, AAS standard solution, Specpure™ As 1000μg/mL, Thermo Scientific Chemicals
Chemical Name or MaterialArsenic AAS standard solution
ConcentrationAs in 5% HNO3
Name NoteAs 1000μg/mL
Health Hazard 1H290-H314-H335-H350
Quantity500 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.