Thermo Scientific Chemicals

Dicyclohexylchlorophosphine, 98+%, Thermo Scientific Chemicals

Catalog number: 033571.03
1 g, Each
Thermo Scientific Chemicals

Dicyclohexylchlorophosphine, 98+%, Thermo Scientific Chemicals

Catalog number: 033571.03
1 g, Each
Quantity
Catalog number: 033571.03
also known as 033571-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16523-54-9
IUPAC Name
chlorodicyclohexylphosphane
Molecular Formula
C12H22ClP
InChI Key
AKJFBIZAEPTXIL-UHFFFAOYSA-N
SMILES
ClP(C1CCCCC1)C1CCCCC1
Form
Liquid
Appearance (Color)
Colorless
Assay (unspecified)
> 98.0%

Description

Dicyclohexylchlorophosphine is used as pharmaceutical intermediate. It is also used in the synthesis of various phosphines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text