Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer, 99%, Thermo Scientific Chemicals
Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 033657.MD
also known as 033657-MD
Price (USD)/ Each
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS12354-85-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to brown
Elemental AnalysisHydrogen: 4.9 ? 0.4%
Infrared spectrumConforms
FormCrystalline solid
Assay (unspecified)33.3 ? 0.7% (Rhodium)
View more
Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer is used as a catalyst for amidation, reductive alkylation, hydrogenation, chiral hydrogenation reactions. It is also used as a catalyst for oxidative olefination reactions, C-C bond cleavage of secondary alcohols, oxidative annulation of pyridine and ortho C-H olefination of phenol derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only