(2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane, 99%, Thermo Scientific Chemicals
(2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
250 mg
Catalog number 033668.MD
also known as 033668-MD
Price (USD)/ Each
94.70
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
94.70
Add to cart
(2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane, 99%, Thermo Scientific Chemicals
Catalog number033668.MD
Price (USD)/ Each
94.70
-
Add to cart
Chemical Identifiers
CAS77876-39-2
IUPAC Name[(2S,4S)-4-(diphenylphosphanyl)pentan-2-yl]diphenylphosphane
Molecular FormulaC29H30P2
InChI KeyCTYPJIUQROQJBG-ISIPKWIHNA-N
SMILESC[C@@H](C[C@H](C)P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥98.5%
Phosphorus-31 NMRConforms to structure
Enantiomeric excess≥98.5%
Proton NMRConforms to structure
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only