Thermo Scientific Chemicals

Trimethylgermanium chloride, Thermo Scientific Chemicals

Catalog number: 034129.03
1 g, Each
Thermo Scientific Chemicals

Trimethylgermanium chloride, Thermo Scientific Chemicals

Catalog number: 034129.03
1 g, Each
Quantity
Catalog number: 034129.03
also known as 034129-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1529-47-1
Molecular Formula
C3H11ClGe
InChI Key
TWBCUFVBXMWAQL-UHFFFAOYSA-N
SMILES
Cl.C[GeH](C)C
Molecular Weight (g/mol)
155.20
Form
Liquid
Appearance (Color)
Colorless
Assay (unspecified)
≥97.0%

Description

Trimethylgermanium chloride is used to prepare trimethyl(2-thienyl)germane. It is also used as alkylating and precursor to prepare Grignard reagent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text