Thermo Scientific™
Omni View Online Elemental Analyzer Interface Software
Catalog number: 035050-102
Have Questions?
Thermo Scientific™
Omni View Online Elemental Analyzer Interface Software
Catalog number: 035050-102
Have Questions?
The Thermo Scientific™ Omni View GUI is a premier GUI for bulk process equipment, providing a flexible, highly configurable interface for our online elemental analyzers, allowing users and specific industries to optimally process, display, and archive data to meet individual requirements. At the same time, it tracks instrument and system health providing insights to minimize downtime and instigate timely repair. With a multi-year database and extensive capabilities for trend and report generation, material and product tracking, Omni View can be simultaneously accessed and individually configured by multiple users through any number of networked computer systems. Maximize the value of online elemental analysis with Omni View to deliver information effectively, to everyone that needs it, where and when they want it, in a format of their choice.
Catalog Number
035050-102
Unit Size
Each
Price (USD)
Full specifications
DescriptionHighly configurable online elemental analyzer interface software to process, display and archive data
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizePrice (USD)
035050-102Full specifications
EachRequest A Quote
DescriptionHighly configurable online elemental analyzer interface software to process, display and archive data
Unit SizeEach
Showing 1 of 1
Omni View is the product of over three decades of industrial experience and benefits from extensive user input, making it a highly refined solution for maximizing the value and utility of online elemental analyses. Key features include:
  • Highly configurable with respect to, for example, screen layout, content, data density, color, and appearance, for customization to support efficient information flow for every user and industry.
  • Comprehensive charting and trending functionality for optimum data display.
  • Multi-workstation capabilities, for access via remote workstations across the network/plant, wherever required.
  • Auto Diagnostics for monitoring, recording, and analyzing system health to maximize analyzer uptime.
  • Compatible with most major communication protocols for easy integration with existing information and control networks.
  • Connectivity to local service center for rapid, interactive remote assistance.
  • Options for integration with additional software packages such as RAMOS, COBOS, PREBOS for industry-specific optimization.