Sodium borodeuteride, 99% (Isotopic), Thermo Scientific Chemicals
Sodium borodeuteride, 99% (Isotopic), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium borodeuteride, 99% (Isotopic), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 035102.06
Price (USD)/ Each
274.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
323.00 
Save 48.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
274.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
323.00 
Save 48.35 (15%)
Add to cart
Sodium borodeuteride, 99% (Isotopic), Thermo Scientific Chemicals
Catalog number035102.06
Price (USD)/ Each
274.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
323.00 
Save 48.35 (15%)
-
Add to cart
Chemical Identifiers
CAS15681-89-7
IUPAC Namesodium boranuide
Molecular FormulaBH4Na
InChI KeyYOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES[BH4-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormPowder
Proton NMRfor Chemical Purity: Conforms
Proton NMRfor Isotopic Enrichment: ≥99.0%
Purity≥95.0% (titration)
Sodium borodeuteride is used to prepare alfa, alfa-dideutero-beta-amino acids. It is also used to prepare amine-stabilized, deuterated borane-tetrhydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only