Thermo Scientific Chemicals
Interference Check Standard A(ICS-A) Solution, Specpure™, Thermo Scientific Chemicals
Catalog number: 035256.AP
500 mL, Each
Thermo Scientific Chemicals
Interference Check Standard A(ICS-A) Solution, Specpure™, Thermo Scientific Chemicals
Catalog number: 035256.AP
500 mL, Each
Quantity
Catalog number: 035256.AP
also known as 035256-AP
Price (USD)
Quantity
-
Have Questions?
Concentration
Al 5,000?g/mL ?2%
Concentration
Fe 2,000?g/mL ?2%
Concentration
Ca 5,000?g/mL ?2%
Concentration
Mg 5,000?g/mL ?2%