This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 035422.AE
also known as 035422-AE
Price (USD)/ Each
148.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
175.00
Save 26.35 (15%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
148.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
175.00
Save 26.35 (15%)
Add to cart
Water Spiking Standard A (WSS-A) Solution, Specpure™, Thermo Scientific Chemicals
Chemical Name or MaterialWater Spiking Standard A (WSS-A) Solution
ConcentrationMatrix: 5% HNO3
Health Hazard 1H290-H314-H335
Quantity100 mL
Health Hazard 2GHS H Statement H314 Causes severe skin burns and eye damage.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.