Thermo Scientific Chemicals

Magnesium oxide, ACS, 95.0% min, Thermo Scientific Chemicals

Catalog number: 035442.36
500 g, Each
Thermo Scientific Chemicals

Magnesium oxide, ACS, 95.0% min, Thermo Scientific Chemicals

Catalog number: 035442.36
500 g, Each
Quantity
Catalog number: 035442.36
also known as 035442-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1309-48-4
IUPAC Name
oxomagnesium
Molecular Formula
MgO
InChI Key
CPLXHLVBOLITMK-UHFFFAOYSA-N
SMILES
O=[Mg]
Impurity content
NO₃ 0.005%
Impurity content
Sr 0.005%
Assay (unspecified)
95.0% MgO min
Impurity content
Ba 0.005%
Impurity content
Cl 0.01%

Description

Magnesium oxide is used as a refractive material, which is physically and chemically stable at higher temperature. Magnesium oxide is a raw material for producing portland cement. It is also used in agricultural, chemical construction, environment al and other industrial applications. It is also used in medicine, colorimetry, industrial cables, heating elements, ceramics, spin-tunneling devices, and several other materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text