Chemical Identifiers
CAS75965-35-4
IUPAC Namelithium(1+) tris(1,2-dimethoxyethane) tetraphenylboranuide
Molecular FormulaC36H50BLiO6
InChI KeyADYUZFWVPWDPFK-UHFFFAOYSA-N
SMILES[Li+].COCCOC.COCCOC.COCCOC.C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more