Sodium aluminum oxide, tech., Thermo Scientific Chemicals
Sodium aluminum oxide, tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium aluminum oxide, tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 035453.36
also known as 035453-36
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
Add to cart
Sodium aluminum oxide, tech., Thermo Scientific Chemicals
Catalog number035453.36
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1302-42-7
IUPAC Namesodium dioxoalumanuide
Molecular FormulaAlNaO2
InChI KeyIYJYQHRNMMNLRH-UHFFFAOYSA-N
SMILES[Na+].O=[Al-]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble material1% max
Sodium aluminum oxide is used in the isomerization of alkene and isomerization of amines. It is also used in water treatment as an adjunct and as a coagulant aid to flocculation. It is involved in the removal of silica and phosphates from waste water. It is employed to accelerate concrete solidification. It finds application in the paper industry and fire brick production. It acts as an intermediate in the production of zeolites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only