Lithium nitride, 99.4% (metals basis), Thermo Scientific Chemicals
Lithium nitride, 99.4% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium nitride, 99.4% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 035479.18
also known as 035479-18
Price (USD)/ Each
263.65
Online exclusive
293.00 
Save 29.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
263.65
Online exclusive
293.00 
Save 29.35 (10%)
Add to cart
Lithium nitride, 99.4% (metals basis), Thermo Scientific Chemicals
Catalog number035479.18
Price (USD)/ Each
263.65
Online exclusive
293.00 
Save 29.35 (10%)
-
Add to cart
Chemical Identifiers
CAS26134-62-3
IUPAC NameLithium nitride
Molecular FormulaLi3N
InChI KeyIDBFBDSKYCUNPW-UHFFFAOYSA-N
SMILES[Li]N([Li])[Li]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.6% max.
Lithium nitride is used in metallurgy and chemical synthesis. It is also used to store hydrogen and acts as a source of the nitride ion. It is involved in the preparation of lithium hydride and lithium amide as well as a reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only