Thermo Scientific Chemicals
Nitrate, Ion chromatography standard solution, Specpure™ NO{3}∣- 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 035549.AP
500 mL, Each
Thermo Scientific Chemicals
Nitrate, Ion chromatography standard solution, Specpure™ NO{3}∣- 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 035549.AP
500 mL, Each
Quantity
Catalog number: 035549.AP
also known as 035549-AP
Price (USD)
Quantity
-
Have Questions?
Concentration
1,000?g/mL ?1%
Concentration
Nitrate 1000?g/ml
Starting Material
NaNO3
Description
Nitrate, Ion chromatography standard solution is used as a standard solution in analytical chemistry and in ion chromatography. It is used for calibration of ion chromatography and other analytical techniques.